missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butyne, 98%

Product Code. 11407886
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407886 5 g 5g
11417886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11407886 Supplier Thermo Scientific Alfa Aesar Supplier No. A14013.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 917-92-0 | C6H10 | 82.146 g/mol

3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

Solubility
Miscible with chloroform, benzene and toluene. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 917-92-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008852
InChI Key PPWNCLVNXGCGAF-UHFFFAOYSA-N
PubChem CID 13512
IUPAC Name 3,3-dimethylbut-1-yne
SMILES CC(C)(C)C#C

Spécification

Melting Point -78°C
Density 0.671
Boiling Point 37°C to 38°C
Flash Point <−34°C (−29°F)
Odor Recognizable
Linear Formula (CH3)3CC≡CH
Refractive Index 1.3745
Quantity 5 g
UN Number UN3295
Beilstein 1697100
Synonym 3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
Solubility Information Miscible with chloroform,benzene and toluene. Immiscible with water.
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 3,3-Dimethyl-1-butyne
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.