missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, 95%

Product Code. 15595216 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595216 5 g 5g
15505226 250 mg 250mg
15340208 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15595216 Supplier Thermo Scientific Acros Supplier No. 456900050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 887144-97-0 | C10H10F3IO | 330.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887144-97-0
Molecular Formula C10H10F3IO
Molecular Weight (g/mol) 330.09
InChI Key HVAPLSNCVYXFDQ-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi
PubChem CID 16043572
IUPAC Name 3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
SMILES CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C

Specifications

Melting Point 75.0°C to 79.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 330.09
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.