missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylacryloyl chloride, 97%

Product Code. 10153860 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10153860

Brand: Thermo Scientific Acros 179730250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3350-78-5 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3350-78-5
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00000730
InChI Key BDUBTLFQHNYXPC-UHFFFAOYSA-N
Synonym 3,3-dimethylacryloyl chloride, 3-methylcrotonoyl chloride, 3-methyl-but-2-enoyl chloride, 3,3-dimethylacrylyl chloride, 2-butenoyl chloride, 3-methyl, 3-methyl-2-butenoyl chloride, 3,3-dimethylacryloylchloride, dimethylacrylyl chloride, dimethyl acryloyl chloride, acmc-1cpb0
PubChem CID 102394
IUPAC Name 3-methylbut-2-enoyl chloride
SMILES CC(C)=CC(Cl)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 145.0°C to 147.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCOCl
Refractive Index 1.4750 to 1.4780
Quantity 25 g
Beilstein 02, 433
Specific Gravity 1.06
Solubility Information Solubility in water: reacts
Formula Weight 118.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylacryloyl chloride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethylacryloyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.