missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals

Product Code. 11473198
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473198 50 g 50g
11483198 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473198 Supplier Thermo Scientific Alfa Aesar Supplier No. A18958.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-16-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00036569
InChI Key XXIKYCPRDXIMQM-UHFFFAOYSA-N
Synonym prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate
PubChem CID 14489
IUPAC Name 3-methylbut-2-enyl acetate
SMILES CC(C)=CCOC(C)=O

Specifications

Density 0.915
Boiling Point 140°C
Flash Point 40°C (104°F)
Odor Fruit-like
Refractive Index 1.43
Quantity 50 g
UN Number UN3272
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 3,3-Dimethylallyl acetate, stabilized with 0.1% alpha tocopherol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.