missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]benzaldehyde, 95%, Thermo Scientific™

Product Code. 10422834
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10422834 250 mg Amber Glass Bottle 250mg
10652885 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422834 Supplier Thermo Scientific Maybridge Supplier No. CC59404CB
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26815-13-4
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD09064988
InChI Key CMVPGTWBRBJOQR-UHFFFAOYSA-N
Synonym 3-3-dimethylamino propoxy benzaldehyde, benzaldehyde,3-3-dimethylamino propoxy, 3-3-dimethylamino-propoxy-benzaldehyde, 3-3-dimethylaminopropoxy benzaldehyde
PubChem CID 10727031
IUPAC Name 3-[3-(dimethylamino)propoxy]benzaldehyde
SMILES CN(C)CCCOC1=CC=CC(=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 207.27
Percent Purity 95%
Chemical Name or Material 3-[3-(Dimethylamino)propoxy]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.