missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric anhydride, 97%

Product Code. 11420868
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420868 25 g 25g
11410868 5 g 5g
11430868 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420868 Supplier Thermo Scientific Alfa Aesar Supplier No. B25286.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4160-82-1 | C7H10O3 | 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4160-82-1
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00006684
InChI Key HIJQFTSZBHDYKW-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric anhydride, 2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl, 4,4-dimethyldihydro-2h-pyran-2,6 3h-dione, glutaric anhydride, 3,3-dimethyl, 3,3-dimethylglutaricanhydride, glutaric anhydride, beta,beta-dimethyl, acmc-1adve, dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione, 3,3-dimethyladipic anhydride, 3,3 dimethylglutaric anhydride
PubChem CID 77813
IUPAC Name 4,4-dimethyloxane-2,6-dione
SMILES CC1(CC(=O)OC(=O)C1)C

Specifications

Melting Point 122°C to 126°C
Boiling Point 181°C (25 mmHg)
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material 3,3-Dimethylglutaric anhydride
compliance-icons
Product Identifier
  • 3,3-Dimethylglutaric anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3,3-Dimethylglutaricanhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.