missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutarimide, 99%

Product Code. 11455816
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455816 25 g 25g
11449737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455816 Supplier Thermo Scientific Alfa Aesar Supplier No. B22196.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1123-40-6 | C7H11NO2 | 141.17 g/mol

3,3'-dimethylglutarimide were used as the barbital subsitutes. 3,3-di­methyl­glutarimide is involved formation of the heterodimer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-dimethylglutarimide were used as the barbital subsitutes. 3,3-di­methyl­glutarimide is involved formation of the heterodimer.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-40-6
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00006671
InChI Key YUJCWMGBRDBPDL-UHFFFAOYSA-N
Synonym 3,3-dimethylglutarimide, 2,6-piperidinedione, 4,4-dimethyl, glutarimide, 3,3-dimethyl, 4,4-dimethyl-2,6-piperidinedione, beta,beta-dimethylglutarimide, 3,3-dimethyl-glutarimide, .beta.,.beta.-dimethylglutarimide, 4,6-piperidinedione, 2, 4,4-dimethyl, glutarimide,3-dimethyl
PubChem CID 14292
IUPAC Name 4,4-dimethylpiperidine-2,6-dione
SMILES CC1(CC(=O)NC(=O)C1)C

Specifications

Melting Point 144°C to 146°C
Quantity 25 g
Beilstein 111981
Solubility Information Soluble in water.
Formula Weight 141.17
Percent Purity 99%
Chemical Name or Material 3,3-Dimethylglutarimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.