missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dithiodipropionic acid, 98%

Product Code. 10726191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10726191 250 g Glass bottle 250g
10716191 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10726191 Supplier Thermo Scientific Acros Supplier No. 148902500
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-62-6
Molecular Formula C6H10O4S2
Molecular Weight (g/mol) 210.26
MDL Number MFCD00002780
InChI Key YCLSOMLVSHPPFV-UHFFFAOYSA-N
Synonym 3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid
PubChem CID 95116
IUPAC Name 3-(2-carboxyethyldisulfanyl)propanoic acid
SMILES C(CSSCCC(=O)O)C(=O)O

Specifications

CAS Min % 98
Melting Point 155°C to 158°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula S2(CH2CH2CO2H)2
Quantity 250 g
Beilstein 03, IV, 736
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,benzene
Formula Weight 210.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 3'-Dithiodipropionic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3'-Dithiodipropionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.