missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10225934
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225934 250 mg Amber Glass Bottle 250mg
10797665 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225934 Supplier Thermo Scientific Maybridge Supplier No. CC49504CB
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 273727-50-7
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD09817468
InChI Key YUNJEYQSCYYAEF-UHFFFAOYSA-N
Synonym 3-3-methyl-1,2,4-oxadiazol-5-yl benzaldehyde, benzaldehyde,3-3-methyl-1,2,4-oxadiazol-5-yl, 3-3-methyl-1,2,4 oxadiazol-5-yl-benzaldehyde
PubChem CID 18545510
IUPAC Name 3-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
SMILES CC1=NOC(=N1)C1=CC(C=O)=CC=C1

Especificaciones

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 3-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzaldehyde
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.