missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Tolidine, TRC

Product Code. 30455279
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455279 100 g 100g
30450832 50 g 50g
30453564 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455279 Supplier Toronto Research Chemicals Supplier No. TRCT535330100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-93-7
Molecular Formula C14 H16 N2
Molecular Weight (g/mol) 212.29
Synonym Benzidine, 3,3'-dimethyl- (8CI), 3,3'-Dimethyl[1,1'-biphenyl]-4,4'-diamine, 3,3'-Dimethyl-4,4'-biphenyldiamine, 3,3'-Dimethyl-4,4'-diaminobiphenyl, 3,3'-Dimethylbenzidine, 3,3'-Tolidine, 4,4'-Diamino-3,3'-dimethyl-1,1'-biphenyl, 4,4'-Diamino-3,3'-dimethylbiphenyl, 4'-Amino-3,3'-dimethyl[1,1'-biphenyl]-4-ylamine, C.I. 37230, C.I. Azoic Diazo Component 113, Fast Dark Blue Base R, o,o'-Tolidine, o-Tolidine
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
SMILES Cc1cc(ccc1N)c2ccc(N)c(C)c2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C14H16N2/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12/h3-8H,15-16H2,1-2H3
Formula Weight 212.1313
Chemical Name or Material 3,3'-Dimethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.