missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Dihydrochloride, TRC

Product Code. 30357655
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357655 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357655 Supplier Toronto Research Chemicals Supplier No. TRCC37949550MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Dihydrochloride
CAS 2512213-25-9
Molecular Formula C17H29Cl3N2O
Quantity 50 mg
Synonym Trazodone Impurity G
InChI Formula InChI=1S/C17H27ClN2O.2ClH/c1-15(2)14-21-12-4-7-19-8-10-20(11-9-19)17-6-3-5-16(18)13-17;;/h3,5-6,13,15H,4,7-12,14H2,1-2H3;2*1H
SMILES CC(C)COCCCN1CCN(C2=CC=CC(Cl)=C2)CC1.[H]Cl.[H]Cl
IUPAC Name 1-(3-chlorophenyl)-4-(3-isobutoxypropyl)piperazine dihydrochloride
Molecular Weight (g/mol) 383.78
Formula Weight 382.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.