missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4-Dimethyl-2-oxazolinyl)pyridine, 97%

Product Code. 15456897
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456897 10 g 10g
15446897 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456897 Supplier Thermo Scientific Alfa Aesar Supplier No. H33997.09
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68981-86-2 | C10H12N2O | 176.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68981-86-2
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.219
MDL Number MFCD01313220
InChI Key XYABHURTMBEOGW-UHFFFAOYSA-N
Synonym 4,5-dihydro-4,4-dimethyl-2-3-pyridyl oxazole, 3-4,4-dimethyl-4,5-dihydro-1,3-oxazol-2-yl pyridine, 4,5-dihydro-4,4-dimethyl-2-pyrid-3-yl oxazole, 4,4-dimethyl-2-pyridin-3-yl-4,5-dihydrooxazole, 3-4,4-dimethyl-5h-1,3-oxazol-2-yl pyridine, 4,4-dimethyl-2-3-pyridyl-1,3-oxazoline, pubchem8698, 3-4,4-dimethyl-2-oxazolinyl pyridine, 4,4-dimethyl-2-3-pyridyl-oxazoline, 4,4-dimethyl-2-3-pyridinyl-5h-oxazole
PubChem CID 2736129
IUPAC Name 4,4-dimethyl-2-pyridin-3-yl-5H-1,3-oxazole
SMILES CC1(COC(=N1)C2=CN=CC=C2)C

Specifications

Melting Point 25°C to 27°C
Quantity 10 g
Sensitivity Air Sensitive
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 3-(4,4-Dimethyl-2-oxazolinyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.