missing translation for 'onlineSavingsMsg'
Learn More

3,4,4'-Trichlorocarbanilide(Triclocarban), TRC

Product Code. 30451364
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451364 1 g 1g
30449126 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451364 Supplier Toronto Research Chemicals Supplier No. TRCT7742001G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-20-2
Molecular Formula C13 H9 Cl3 N2 O
Molecular Weight (g/mol) 315.58
Synonym Urea, N-(4-chlorophenyl)-N'-(3,4-dichlorophenyl)-, Carbanilide, 3,4,4'-trichloro- (6CI,7CI,8CI), N-(4-Chlorophenyl)-N'-(3,4-dichlorophenyl)urea, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl) urea, 3,4,4'-Trichlorocarbanilide, 3,4,4'-Trichlorodiphenylurea, 3-(4-Chlorophenyl)-1-(3,4-dichlorophenyl)urea, Cusiter, Cuticura, Cutisan, ENT 26925, Genoface, N-(3,4-Dichlorophenyl)-N'-(4-chlorophenyl)urea, NSC 72005, Preventol SB, Preventol SB extra, Procutene, Solubacter, TCC, Trichlorocarban, Triclocarban
IUPAC Name 1-(4-chlorophenyl)-3-(3,4-dichlorophenyl)urea
SMILES Clc1ccc(NC(=O)Nc2ccc(Cl)c(Cl)c2)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H9Cl3N2O/c14-8-1-3-9(4-2-8)17-13(19)18-10-5-6-11(15)12(16)7-10/h1-7H,(H2,17,18,19)
Formula Weight 313.978
Percent Purity >95%
Chemical Name or Material Triclocarban
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.