missing translation for 'onlineSavingsMsg'
Learn More

3,4,5,6-Tetrahydrophthalic anhydride, 98%

Product Code. 10762433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762433 5 g Glass bottle 5g
10583491 25 g Glass bottle 25g
11905041 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10762433 Supplier Thermo Scientific Acros Supplier No. 138220050
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2426-02-0 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2426-02-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005917
InChI Key HMMBJOWWRLZEMI-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione
PubChem CID 12475
SMILES O=C1OC(=O)C2=C1CCCC2

Specifications

Melting Point 70.0°C to 74.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 17,461
Solubility Information Solubility in water: reacts.
IUPAC Name 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
Formula Weight 152.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5,6-Tetrahydrophthalic anhydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4,5,6-Tetrahydrophthalic anhydride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.