missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trichloropyridazine, 98%, Thermo Scientific Chemicals

Product Code. 15585866 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585866 5 g 5g
15575866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15585866 Supplier Thermo Scientific Acros Supplier No. 160590050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14161-11-6
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.42
InChI Key GBAOOJAWDCNOGO-UHFFFAOYSA-N
Synonym pyridazine, 3,4,5-trichloro, 3,4,5-trichloro-pyridazine, trichloropyridazine, ksc493s5t, gbaoojawdcnogo-uhfffaoysa, pyridazine,3,4,5-trichloro, 3,4,5-trichloro-1,2-diazine
PubChem CID 70111
IUPAC Name 3,4,5-trichloropyridazine
SMILES C1=C(C(=C(N=N1)Cl)Cl)Cl

Specifications

Melting Point 54.0°C to 61.0°C
Boiling Point 117.0°C to 118.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 183.42
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridazine
compliance-icons
Product Identifier
  • 3,4,5-Trichloropyridazine, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.