missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzoic acid, 98%

Product Code. 11451347
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451347 250 g 250g
11441347 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451347 Supplier Thermo Scientific Alfa Aesar Supplier No. B24887.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149-91-7 | C7H6O5 | 170.12 g/mol

Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

Solubility
Soluble in water, alcohol, ether, glycerol and acetone. Insoluble in benzene, chloroform and petroleum ether.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents, strong bases, acid anhydrides, acid chlorides, metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002510
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
PubChem CID 370
ChEBI CHEBI:30778
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point ∼251°C (decomposition)
Density 1.694
Quantity 250 g
Beilstein 2050274
Sensitivity Hygroscopic; Light sensitive
Merck Index 144345
Solubility Information Soluble in water,alcohol,ether,glycerol and acetone. Insoluble in benzene,chloroform and petroleum ether.
Formula Weight 170.12
Percent Purity 0.99
Chemical Name or Material 3,4,5-Trihydroxybenzoic acid, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.