missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzoic acid monohydrate, ACS, 98+%

Product Code. 11377878
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377878 500 g 500g
11387878 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377878 Supplier Thermo Scientific Alfa Aesar Supplier No. 036417.36
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol

3,4,5-Trihydroxybenzoic acid monohydrate is used in making dyes and inks, photographic developers and has been used as astringents in medically. It is also are used as antioxidants in foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 250°C (decomposition)
Color White to Cream
Quantity 500 g
Sensitivity Light sensitive
Merck Index 144345
Solubility Information Soluble in water(1.1g/100ml).
IUPAC Name 3,4,5-trihydroxybenzoic acid;hydrate
Formula Weight 188.14 (170.12 Anhydrous)
Percent Purity ≥98%
Grade ACS
Physical Form Powder
Chemical Name or Material 3,4,5-Trihydroxybenzoic acid monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.