missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzoic Acid, Pure, Fisher Chemical

Product Code. 10040510 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10040510 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10040510 Supplier Fisher Chemical Supplier No. T/3300/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 149-91-7
Melting Point 253°C
Color Yellow
Molecular Formula C7H6O5
MDL Number MFCD00002510
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1
IUPAC Name 3,4,5-trihydroxybenzoic acid
Molecular Weight (g/mol) 170.12
PubChem CID 370
ChEBI CHEBI:30778
Formula Weight 170.12g/mol
Quantity 100 g
Packaging Plastic powder jar
Physical Form Powder Solid
Show More Show Less
compliance-icons
Product Identifier
  • Gallic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.