missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzeneboronic acid, 98%

Product Code. 11398186
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398186 1 g 1g
11308196 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398186 Supplier Thermo Scientific Alfa Aesar Supplier No. L15191.03
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 182163-96-8 | C9H13BO5 | 212.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182163-96-8
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD01075676
InChI Key RULQUTYJXDLRFL-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
PubChem CID 2734390
IUPAC Name (3,4,5-trimethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O

Specifications

Melting Point ∼245°C
Quantity 1 g
Beilstein 8205051
Formula Weight 212.01
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.