missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxycinnamic acid, MedChemExpress

Product Code. 16449356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449356 Supplier Medchem Express Supplier No. HYW012123

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3,4,5-Trimethoxycinnamic acid is a phenylpropanoid isolated from the roots of Polygala tenuifolia WILLD, with anti-stress effect, prolonging the sleeping time in animals. 3,4,5-Trimethoxycinnamic acid increases expression of GAD65 and γ-subunit of GABAA receptor, but shows no effect on the amounts of α-, β-subunits.

3,4,5-Trimethoxycinnamic acid is a phenylpropanoid isolated from the roots of Polygala tenuifolia

Specifications

Chemical Name or Material 3,4,5-Trimethoxycinnamic acid
CAS 90-50-6
Color White
Purity Grade Notes Research
Molecular Formula C12H14O5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (419.74 mM; Need ultrasonic)
SMILES COC1=C(OC)C(OC)=CC(/C=C/C(O)=O)=C1
Molecular Weight (g/mol) 238.24
Formula Weight 238.24
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.