missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenol, TRC

Product Code. 30451115
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451115 Supplier Toronto Research Chemicals Supplier No. TRCT7958951G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4,5-Trimethoxyphenol
CAS 642-71-7
Molecular Formula C13 H24 N2 O
Quantity 1 g
Synonym 3,4,5-Trimethoxy-phenol, 3',4',5'-Trimethoxyphenol, Antiarol
InChI Formula InChI=1S/C13H24N2O/c1-3-5-14-7-11-9-15(6-4-2)10-12(8-14)13(11)16/h11-12H,3-10H2,1-2H3
SMILES CCCN1CC2CN(CCC)CC(C1)C2=O
IUPAC Name 3,7-dipropyl-3,7-diazabicyclo[3.3.1]nonan-9-one
Molecular Weight (g/mol) 224.343
Formula Weight 224.189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.