missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylboronic acid, 97%, ACROS Organics™

Product Code. 10234402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10234402 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10234402 Supplier Thermo Scientific Acros Supplier No. 359770010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 182163-96-8
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H13BO5
Quantity 1 g
Synonym 3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
InChI Key RULQUTYJXDLRFL-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O
IUPAC Name (3,4,5-trimethoxyphenyl)boronic acid
Molecular Weight (g/mol) 212.008
PubChem CID 2734390
Formula Weight 212.01
Chemical Name or Material 3, 4, 5-Trimethoxyphenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.