missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-acetyl-D-glucal, 98%, Thermo Scientific Chemicals

Product Code. 11358975
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
11358975 25 g 25g
11348975 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11358975 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor L14103.14
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 2873-29-2 | Molecular Formula: C12H16O7 | Molecular Weight: 272.253 g/mol

3,4,6-Tri-O-acetyl-D-glucal acts as a building block for the synthesis of oligosaccharides in both solution and solid-phase. Further, it is used in the preparation of D-arabino-1,5-anhydro-2-deoxy-hex-1-enitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-acetyl-D-glucal acts as a building block for the synthesis of oligosaccharides in both solution and solid-phase. Further, it is used in the preparation of D-arabino-1,5-anhydro-2-deoxy-hex-1-enitol.

Solubility
Soluble in chloroform and ethanol. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 2873-29-2
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.253
MDL Number MFCD00063253
InChI Key LLPWGHLVUPBSLP-UTUOFQBUSA-N
Synonym tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
PubChem CID 688303
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C

Especificaciones

Melting Point 52°C to 55°C
Quantity 25 g
Beilstein 90781
Solubility Information Soluble in chloroform and ethanol. Insoluble in water.
Optical Rotation −25° (c=1 in Methanol)
Formula Weight 272.26
Percent Purity 98%
Chemical Name or Material 3,4,6-Tri-O-acetyl-D-glucal

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.