missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione, 99%, BLD Pharm™

Product Code. 30802706
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30802706 1 g 1g
30794135 250 mg 250mg
30795897 100 g 100g
30793697 25 g 25g
30794335 10 g 10g
30794862 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30802706 Supplier BLD Pharm Supplier No. BD1361281g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 191732-72-6
Molecular Formula C13H13N3O3
MDL Number MFCD07772307
Quantity 1 g
SMILES C1=CC=C(C3=C1C(N(C2C(NC(=O)CC2)=O)C3)=O)N
Molecular Weight (g/mol) 259.26
Percent Purity 99%
Chemical Name or Material 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.