missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenyl)propionic acid, 98%

Product Code. 15415487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415487 5 g 5g
15405487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415487 Supplier Thermo Scientific Alfa Aesar Supplier No. H32729.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35888-99-4 | C15H14O2 | 226.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35888-99-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD06823979
InChI Key MVFHRQWYCXYYMU-UHFFFAOYSA-N
Synonym 2 4-biphenyl propionic acid, 3-4-phenylphenyl propanoic acid, 3-4-biphenyl propionic acid, 3-1,1'-biphenyl-4-yl propanoic acid, 3-biphenyl-4-yl propanoic acid, 1,1'-biphenyl-4-propanoic acid, 2 para-biphenyl propionic acid, biphenyl-4-propanoic acid
PubChem CID 3082434
IUPAC Name 3-(4-phenylphenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 149°C to 152°C
Quantity 5 g
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material 3-(4-Biphenyl)propionic acid
compliance-icons
Product Identifier
  • 3-(4-Biphenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: [1,1-Biphenyl]-4-propanoic acid

RUO – Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.