missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)-1,2-propanediol, 99%

Product Code. 11413637
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11413637 500 g 500g
11493627 25 g 25g
11403637 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11413637

Brand: Thermo Scientific Alfa Aesar A17386.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-29-0 | C9H11ClO3 | 202.634 g/mol

It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

Solubility
Slightly soluble in water.

Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-29-0
Molecular Formula C9H11ClO3
Molecular Weight (g/mol) 202.634
MDL Number MFCD00021990
InChI Key MXOAEAUPQDYUQM-UHFFFAOYSA-N
Synonym chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
PubChem CID 7697
ChEBI CHEBI:3642
IUPAC Name 3-(4-chlorophenoxy)propane-1,2-diol
SMILES C1=CC(=CC=C1OCC(CO)O)Cl

Specifications

Melting Point 78°C to 82°C
Quantity 500 g
Solubility Information Slightly soluble in water.
Formula Weight 202.64
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorophenoxy)-1,2-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.