missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)-1,2-propanediol(Chlorphenesin), TRC

Product Code. 30136711
Change view
Click to view available options
Quantity:
2.5 g
25 g
50 g
Unit Size:
2.5g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136711 50 g 50g
30134256 25 g 25g
17902885 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136711 Supplier Toronto Research Chemicals Supplier No. TRCC42425050G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-29-0
Molecular Formula C9 H11 Cl O3
Molecular Weight (g/mol) 202.63
Synonym 1,2-Propanediol, 3-(p-chlorophenoxy)- (6CI,7CI,8CI), 3-(4-Chlorophenoxy)-1,2-propanediol, (±)-Chlorphenesin, (±)-p-Chlorphenesin, 3-(p-Chlorophenoxy)propane-1,2-diol, Adermykon, Chlorphenesin, Demykon, Elestab CPN, Gecophen, Glycerol α-p-chlorophenyl ether, Mycil, NSC 6401, p-Chlorophenyl glyceryl ether, p-Chlorophenyl-α-glyceryl ether, p-Chlorphenesin
IUPAC Name 3-(4-chlorophenoxy)propane-1,2-diol
SMILES OCC(O)COc1ccc(Cl)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H11ClO3/c10-7-1-3-9(4-2-7)13-6-8(12)5-11/h1-4,8,11-12H,5-6H2
Formula Weight 202.0397
Chemical Name or Material Chlorphenesin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.