missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminopyridine, 98%

Product Code. 10346320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10346320 5 g Glass bottle 5g
10542364 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346320 Supplier Thermo Scientific Supplier No. 112390050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 54-96-6 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-96-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006401
InChI Key OYTKINVCDFNREN-UHFFFAOYSA-N
Synonym 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
PubChem CID 5918
IUPAC Name pyridine-3,4-diamine
SMILES C1=CN=CC(=C1N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 216°C to 221°C
Color Brown to Brown-Gray
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(NH2)2
Quantity 5 g
Beilstein 22, II, 395
Merck Index 15, 398
Solubility Information Solubility in water: 24g/L (20°C)
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Diaminopyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Diaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.