Learn More
3,4-Diaminopyridine, 97%
Description
It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.
Solubility
Soluble in water (24 mg/ml at 20°C), alcohol. Slightly soluble in ether.
Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 217°C to 220°C |
| Flash Point | 240°C (464°F) |
| Odor | Unpleasant |
| Quantity | 5 g |
| UN Number | UN2811 |
| Beilstein | 110232 |
| Merck Index | 14,2986 |
| Solubility Information | Soluble in water (24mg/ml at 20°C),alcohol. Slightly soluble in ether. |
| Formula Weight | 109.13 |
| Percent Purity | ≥98% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.