missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminothiophene dihydrochloride, 96%

Product Code. 10315901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10315901 1 g Glass bottle 1g
10389360 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 10315901 Leverandør Thermo Scientific Acros Leverandørnr. 325890010
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 90069-81-1 | C4H6N22HCl | 187.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 90069-81-1
Molecular Formula C4H6N22HCl
Molecular Weight (g/mol) 187.09
MDL Number MFCD00671511
InChI Key RAMOMCXNLLLICQ-UHFFFAOYSA-N
Synonym 3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
PubChem CID 12707342
IUPAC Name thiophene-3,4-diamine;dihydrochloride
SMILES C1=C(C(=CS1)N)N.Cl.Cl

Tekniske data

CAS Min % 95.0
CAS Max % 100.0
Melting Point 240.0°C to 245.0°C
Color Brown or Gray
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 187.09
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Diaminothiophene dihydrochloride, 99%
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.