missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminothiophene dihydrochloride, 96%

Product Code. 10389360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10389360 5 g Glass bottle 5g
10315901 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10389360 Supplier Thermo Scientific Acros Supplier No. 325890050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90069-81-1 | C4H6N2S·2HCl | 187.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90069-81-1
Molecular Formula C4H6N2S·2HCl
Molecular Weight (g/mol) 187.09
MDL Number MFCD00671511
InChI Key RAMOMCXNLLLICQ-UHFFFAOYSA-N
Synonym 3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
PubChem CID 12707342
IUPAC Name thiophene-3,4-diamine;dihydrochloride
SMILES C1=C(C(=CS1)N)N.Cl.Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 240.0°C to 245.0°C
Color Brown or Gray
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 187.09
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Diaminothiophene dihydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.