missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzotrifluoride, TRC

Product Code. 30370095
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370095 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370095 Supplier Toronto Research Chemicals Supplier No. TRCD438808500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 3,4-Dichlorobenzotrifluoride
CAS 328-84-7
Molecular Formula C7H3Cl2F3
Quantity 500 mg
Synonym 1,2-Dichloro-4-(trifluoromethyl)benzene, (3,4-Dichlorophenyl)trifluoromethane, 1-(Trifluoromethyl)-3,4-dichlorobenzene, 3,4-Dichloro(trifluoromethyl)benzene, 3,4-Dichloro-1-(trifluoromethyl)benzene, 3,4-Dichloro-alpha,alpha,alpha-trifluorotoluene, 4-(Trifluoromethyl)-1,2-dichlorobenzene, Oxsol 1000
InChI Formula InChI=1S/C7H3Cl2F3/c8-5-2-1-4(3-6(5)9)7(10,11)12/h1-3H
SMILES FC(F)(F)c1ccc(Cl)c(Cl)c1
IUPAC Name 1,2-dichloro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 215
Formula Weight 213.9564
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.