missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl isocyanate, 98%

Product Code. 10217380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10217380 1 g Glass bottle 1g
10269353 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217380 Supplier Thermo Scientific Acros Supplier No. 319220010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19752-09-1 | C8H5Cl2NO | 202.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19752-09-1
Molecular Formula C8H5Cl2NO
Molecular Weight (g/mol) 202.04
MDL Number MFCD00673063
InChI Key WLNAHQZFPSSVTP-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl isocyanate, 3,4-dichlorobenzylisocyanate, 1,2-dichloro-4-isocyanatomethyl benzene, benzene, 1,2-dichloro-4-isocyanatomethyl, 1,2-dichloro-4-isocyanatomethyl-benzene, 3,4-dichlorophenyl methanisocyanate, zlchem 123, acmc-20anfi, pubchem2096, 3,4-dichloro-benzyl isocyanate
PubChem CID 2733368
IUPAC Name 1,2-dichloro-4-(isocyanatomethyl)benzene
SMILES C1=CC(=C(C=C1CN=C=O)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.3750g/mL
Boiling Point 282°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5656 to 1.5676
Quantity 1 g
Specific Gravity 1.375
Formula Weight 202.04
Percent Purity 98%
Physical Form Slightly Viscous Liquid
Chemical Name or Material 3, 4-Dichlorobenzyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.