missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl mercaptan, 97%

Product Code. 11372066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372066 5 g 5g
11362066 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372066 Supplier Thermo Scientific Alfa Aesar Supplier No. L03226.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 36480-40-7 | C7H6Cl2S | 193.085 g/mol

Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36480-40-7
Molecular Formula C7H6Cl2S
Molecular Weight (g/mol) 193.085
MDL Number MFCD00039651
InChI Key CEGBRSQPRQXALB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl mercaptan, 3,4-dichlorobenzenemethanethiol, 3,4-dichlorophenyl methanethiol, 3,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 3,4-dichloro, acmc-1aex5, d0z0bb, 3,4-dichlorobenzylmercaptan, 3,4-dichloro-alpha-toluenethiol, 3,4-dichloro-phenyl-methanethiol
PubChem CID 118986
IUPAC Name (3,4-dichlorophenyl)methanethiol
SMILES C1=CC(=C(C=C1CS)Cl)Cl

Specifications

Density 1.362
Boiling Point 152°C to 153°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.607
Quantity 5 g
UN Number UN2810
Beilstein 3237473
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 193.1
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzyl mercaptan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.