missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenyl isothiocyanate, 97%

Product Code. 11499506
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499506 25 g 25g
11489506 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499506 Supplier Thermo Scientific Alfa Aesar Supplier No. A19236.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6590-94-9 | C7H3Cl2NS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6590-94-9
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00037828
InChI Key OSBIEFWIIINTNJ-UHFFFAOYSA-N
Synonym 3,4-dichlorophenyl isothiocyanate, benzene, 1,2-dichloro-4-isothiocyanato, 3,4-dichlorophenylisothiocyanate, benzene, 1,2-dichloro-4-isothiocyanato-9ci, isothiocyanic acid, 3,4-dichlorophenyl ester, 1,2-dichloro-4-isothiocyanato-benzene, acmc-20anfc, 4-12-00-01271 beilstein handbook reference, 3,4-dichloro-1-isothiocyanato-benzene, #
PubChem CID 23065
IUPAC Name 1,2-dichloro-4-isothiocyanatobenzene
SMILES ClC1=CC=C(C=C1Cl)N=C=S

Specifications

Density 1.422
Boiling Point 145°C to 148°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.68
Quantity 25 g
UN Number UN2922
Beilstein 511101
Sensitivity Moisture sensitive
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.