missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylmagnesium bromide, 0.50M in 2-MeTHF, Thermo Scientific™

Product Code. 15451406
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451406 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15451406 Supplier Thermo Scientific Alfa Aesar Supplier No. H54621.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79175-35-2
Quantity 100 mL
Molecular Formula C6H3BrCl2Mg
MDL Number MFCD01319891
UN Number UN2924
Synonym 3,4-dichlorophenylmagnesium bromide, bromo 3,4-dichlorophenyl magnesium, 3,4-dichlorophenylmagnesium bromide solution, 3,4-dichlorphenylmagnesiumbromid, irrosodeqhizev-uhfffaoysa-m, 3,4-dichlorophenyl magnesium bromide, 3,4-dichlorophenyl-magnesium bromide, 3,4-dichlorophenylmagnesium bromide, 0.50m in 2-methf, 3,4-dichlorophenylmagnesium bromide 0.5 m in tetrahydrofuran, grignard reagent
InChI Key IRROSODEQHIZEV-UHFFFAOYSA-M
SMILES ClC1=CC=C([Mg]Br)C=C1Cl
IUPAC Name magnesium;1,2-dichlorobenzene-5-ide;bromide
Molecular Weight (g/mol) 250.20
PubChem CID 11043147
Formula Weight 250.2
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3,4-Dichlorophenylmagnesium bromide
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.