missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diethoxy-3-cyclobutene-1,2-dione, 98%

Product Code. 11446380
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446380 25 g 25g
11426380 1 g 1g
11436380 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446380 Supplier Thermo Scientific Alfa Aesar Supplier No. A19248.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5231-87-8 | C8H10O4 | 170.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5231-87-8
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00001333
InChI Key DFSFLZCLKYZYRD-UHFFFAOYSA-N
Synonym 3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn
PubChem CID 123228
IUPAC Name 3,4-diethoxycyclobut-3-ene-1,2-dione
SMILES CCOC1=C(C(=O)C1=O)OCC

Specifications

Density 1.162
Boiling Point 95°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.509
Quantity 25 g
Beilstein 640626
Formula Weight 170.16
Percent Purity 98%
Chemical Name or Material 3,4-Diethoxy-3-cyclobutene-1,2-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.