missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluoro-benzoic Acid, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 3,4-Difluorobenzoic acid |
| CAS | 455-86-7 |
| Molecular Formula | C7 H4 F2 O2 |
| Quantity | 50 g |
| Synonym | Benzoic acid, 3,4-difluoro- (6CI, 7CI, 8CI, 9CI, ACI), 3,4-Difluorobenzoic acid (ACI), NSC 190686 |
| InChI Formula | InChI=1S/C7H4F2O2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H,(H,10,11) |
| SMILES | OC(=O)c1ccc(F)c(F)c1 |
| IUPAC Name | 3,4-difluorobenzoic acid |
| Molecular Weight (g/mol) | 158.1 |
| Formula Weight | 158.0179 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction