missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzene-1-sulfonyl chloride, 97%, Maybridge

Product Code. 10293712
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10293712 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10293712 Supplier Thermo Scientific Maybridge Supplier No. SPB05941DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 145758-05-0
Density 1.586g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H3ClF2O2S
Linear Formula F2C6H3SO2Cl
Quantity 1 g
Synonym 3,4-difluorobenzene-1-sulfonyl chloride, 3,4-difluorobenzenesulphonyl chloride, 3,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-difluoro, 3,4-difluorophenyl chlorosulfone, 3,4-difluorobenzenesulfonylchloride, 3,4-difluorobenzene sulfonyl chloride, 3,4-difluorobenzenesulphonylchloride, pubchem1958, acmc-1byq8
InChI Key FSGLUBQENACWCC-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)F)F
IUPAC Name 3,4-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol) 212.595
PubChem CID 2736913
Formula Weight 212.6
Percent Purity 97%
Chemical Name or Material 3, 4-Difluorobenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.