missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzothiazine, 97%

Product Code. 15447538
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447538 1 g 1g
15457538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447538 Supplier Thermo Scientific Alfa Aesar Supplier No. H61045.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3080-99-7 | C8H9NS | 151.23 g/mol

3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3080-99-7
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.23
MDL Number MFCD04038593
InChI Key YBBLSBDJIKMXNQ-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 thiazine, 2,3-dihydro-1,4-benzothiazine, 3,4-dihydro-2h-benzo 1,4 thiazine, 2h-1,4-benzothiazine, 3,4-dihydro, benzothiomorpholine, thiaphenomorpholine, 2,3-dihydrobenzo-1,4-thiazine, 2,3-dihydro-4h-1,4-benzothiazine, 2h,3h,4h-benzo e 1,4-thiazine, 2h-1,4-benzothiazine,3,4-dihydro
PubChem CID 151064
IUPAC Name 3,4-dihydro-2H-1,4-benzothiazine
SMILES C1CSC2=CC=CC=C2N1

Specifications

Quantity 1 g
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzothiazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.