missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine, 97%

Product Code. 15436487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436487 5 g 5g
15426487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436487 Supplier Thermo Scientific Alfa Aesar Supplier No. H33641.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5735-53-5 | C8H9NO | 135.166 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5735-53-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD02181098
InChI Key YRLORWPBJZEGBX-UHFFFAOYSA-N
Synonym benzomorpholine, 3,4-dihydro-2h-benzo b 1,4 oxazine, 3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro, 2,3-dihydro-1,4-benzoxazine, akos bc-1149, 3,4-dihydro-2h-1,4-benzoxazin, pubchem22677, acmc-1amxt, 3,4-dihydro-2h-1,4-benzooxazine
PubChem CID 585096
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC=CC=C2N1

Specifications

Boiling Point 85°C (1 mmHg)
Quantity 5 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.