missing translation for 'onlineSavingsMsg'
Learn More

3,4-dihydro-2H-1,4-benzoxazine, Thermo Scientific™

Product Code. 10803011
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10803011 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10803011 Supplier Thermo Scientific Maybridge Supplier No. MAY00300DA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5735-53-5
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H9NO
Quantity 1 g
Synonym benzomorpholine, 3,4-dihydro-2h-benzo b 1,4 oxazine, 3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro, 2,3-dihydro-1,4-benzoxazine, akos bc-1149, 3,4-dihydro-2h-1,4-benzoxazin, pubchem22677, acmc-1amxt, 3,4-dihydro-2h-1,4-benzooxazine
InChI Key YRLORWPBJZEGBX-UHFFFAOYSA-N
SMILES C1COC2=CC=CC=C2N1
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol) 135.166
PubChem CID 585096
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 4-benzoxazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.