missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-amine, 98%

Product Code. 11387469
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387469 1 g 1g
11397469 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387469 Supplier Thermo Scientific Alfa Aesar Supplier No. H27862.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 175136-34-2 | C9H11NO2 | 165.192 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. High soluble in ethanol, acetone.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175136-34-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00218806
InChI Key FVLCICVRAPEYNX-UHFFFAOYSA-N
PubChem CID 2775654
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-amine
SMILES C1COC2=C(C=C(C=C2)N)OC1

Specifications

Melting Point 79°C to 81°C
Boiling Point 123°C to 128°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine
Solubility Information Soluble in water. High soluble in ethanol,acetone.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin-7-amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.