missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-ylboronic acid, 90+%, Thermo Scientific™

Product Code. 10712483
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712483 1 g Amber Glass Bottle 1g
10151112 250 mg Amber Glass Bottle 250mg
10294714 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10712483 Supplier Thermo Scientific Maybridge Supplier No. CC13212DA
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 279261-89-1
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02681910
InChI Key CSNCBPVLUIFJOS-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl boronic acid, 3,4-dihydro-2h-1,5-benzodioxepine-7-boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl boronic acid, boronic acid,b-3,4-dihydro-2h-1,5-benzodioxepin-7-yl, 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-ylboronic acid, boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl, pubchem7845, acmc-1crzt, 3,4-dihydro-2h-1,5-benzodioxepin-7-boronic acid
PubChem CID 2776398
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid
SMILES OB(O)C1=CC=C2OCCCOC2=C1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.99
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.