missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-2699397
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10089091 250 mg Amber Glass Bottle 250mg
10212592 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10089091 Supplier Thermo Scientific Maybridge Supplier No. CC13203CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-38-0
Molecular Formula C9H9ClO4S
Molecular Weight (g/mol) 248.68
MDL Number MFCD02681959
InChI Key ADAGPISZFDLGCR-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-sulfonyl chloride, 2h-1,5-benzodioxepin-7-sulfonylchloride, 3,4-dihydro, 2h-1,5-benzodioxepin-7-sulfonyl chloride, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepan-7-ylchlorosulfone, 3,4-dihydro-2h-1,5-benzodioxepin-7-sulfonyl chloride
PubChem CID 2776386
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(OCCCO2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 248.69
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepine-7-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.