missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-isoquinolin-1-one, 98%

Product Code. 15442242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442242 1 g 1g
15452242 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442242 Supplier Thermo Scientific Acros Supplier No. 458220010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 1196-38-9 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196-38-9
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
InChI Key YWPMKTWUFVOFPL-UHFFFAOYSA-N
Synonym 3,4-dihydroisoquinolin-1 2h-one, 1,2,3,4-tetrahydroisoquinolin-1-one, 1 2h-isoquinolinone, 3,4-dihydro, 3,4-dihydroisocarbostyril, 1-oxo-1,2,3,4-tetrahydroisoquinoline, 3,4-dihydro-1 2h-isoquinolinone, 3,4-dihydro-1h-isoquinolin-1-one, dihydroisoquinolone, 3nuu, 1-keto-1,2,3,4-tetrahydroisoquinoline
PubChem CID 150896
IUPAC Name 3,4-dihydro-2H-isoquinolin-1-one
SMILES C1CNC(=O)C2=CC=CC=C21

Specifications

Melting Point 67.0°C to 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2H-isoquinolin-1-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.