missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran-5-carbaldehyde, Thermo Scientific™

Product Code. 10222402
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Product Code. Quantity unitSize
10222402 1 g 1g
10407744 5 g 5g
10351815 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 10222402

Brand: Thermo Scientific Maybridge KM03842DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25090-33-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00085019
InChI Key WWZVSCJTMPYTLY-UHFFFAOYSA-N
PubChem CID 2736151
IUPAC Name 3,4-dihydro-2H-pyran-5-carbaldehyde
SMILES C1CC(=COC1)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-pyran-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.