missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran-5-carbaldehyde, Thermo Scientific™

Product Code. 10407744
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10407744 5 g 5g
10222402 1 g 1g
10351815 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407744 Supplier Thermo Scientific Maybridge Supplier No. KM03842DE
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 25090-33-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00085019
InChI Key WWZVSCJTMPYTLY-UHFFFAOYSA-N
PubChem CID 2736151
IUPAC Name 3,4-dihydro-2H-pyran-5-carbaldehyde
SMILES C1CC(=COC1)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Synonym 5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-pyran-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.