missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran-5-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 15219716 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15219716 1 g 1g
15229716 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15219716

Brand: Thermo Scientific Acros 450380010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25090-33-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
InChI Key WWZVSCJTMPYTLY-UHFFFAOYSA-N
PubChem CID 2736151
IUPAC Name 3,4-dihydro-2H-pyran-5-carbaldehyde
SMILES C1CC(=COC1)C=O

Specifications

Boiling Point 90.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-pyran-5-carboxaldehyde
compliance-icons
Product Identifier
  • 3,4-Dihydro-2H-pyran-5-carboxaldehyde, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.