missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyrido[1,2-A]pyrimidin-2-one, TRC

Product Code. 30384484
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384484 500 mg -
30369266 50 mg 50mg
30369785 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384484 Supplier Toronto Research Chemicals Supplier No. TRCD493150500MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5439-14-5
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.16
SMILES O=C1CCN2C=CC=CC2=N1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8N2O/c11-8-4-6-10-5-2-1-3-7(10)9-8/h1-3,5H,4,6H2
Formula Weight 148.0637
Chemical Name or Material 3,4-Dihydro-2H-pyrido[1,2-A]pyrimidin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.